Merck
  • Potential role of hematopoietic pre-B-cell leukemia transcription factor-interacting protein in oral carcinogenesis.
Potential role of hematopoietic pre-B-cell leukemia transcription factor-interacting protein in oral carcinogenesis.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology (2014-07-26)
Seiji Okada, Tarou Irié, Junichi Tanaka, Rika Yasuhara, Gou Yamamoto, Tomohide Isobe, Chie Hokazono, Tetsuhiko Tachikawa, Yohko Kohno, Kenji Mishima
ABSTRACT
Hematopoietic pre-B-cell leukemia transcription factor-interacting protein (HPIP) is a corepressor of pre-B-cell leukemia homeobox (PBX) 1 and is known to play a role in hematopoiesis. Recently, HPIP was demonstrated to promote breast cancer cell proliferation and hepatocellular carcinoma growth. Moreover, it has been revealed that homeobox and PBX proteins, the expression of which is regulated by HPIP, play key roles in cancer of various organs, including oral squamous cell carcinoma (OSCC). Nevertheless, there has not been any study regarding the role of HPIP in OSCC. This study investigated the expression of HPIP in normal oral mucosa, epithelial precursor lesion (OEPL), and OSCC, and the functional roles of HPIP in OSCC cells and normal keratinocytes. Immunohistochemical analysis of HPIP, Ki-67, and involucrin was performed in OSCC specimens, and the change in involucrin expression following RNA interference treatment against HPIP was examined by quantitative RT-PCR and Western blot analysis in SCC9 and NHEK cells undergoing extracellular calcium-induced differentiation. Matrigel transwell and cell proliferation assays for both cell lines transfected with HPIP siRNA were also conducted. HPIP expression increased in OEPL and OSCC specimens. In vitro analysis revealed that HPIP suppressed differentiation and proliferation of SCC9 cells and transwell migration of NHEK cells, while HPIP promoted invasion of SCC9 and proliferation of NHEK cells. However, HPIP has no significant effect on NHEK cell differentiation. HPIP may play a critical role in oral carcinogenesis and is thus a potential target for anticancer therapy, with particular emphasis on its involvement in differentiation and migration/metastasis.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)