Merck
  • Surfactant-coated graphitized multiwalled carbon nanotubes as the pseudostationary phase in electrokinetic chromatography for the analysis of phytochemical compounds in biological fluids.
Surfactant-coated graphitized multiwalled carbon nanotubes as the pseudostationary phase in electrokinetic chromatography for the analysis of phytochemical compounds in biological fluids.
Electrophoresis (2015-02-03)
Wan Cao, Shuai-Shuai Hu, Xue-Zhen Ying, Li-Hong Ye, Jun Cao
ABSTRACT
This report describes the use of surfactant-coated graphitized multiwalled carbon nanotubes (SC-GMWNTs) as a novel pseudostationary phase in CE with diode array detection for the determination of phenolic acids and tanshinones in herbal and urine samples. Several parameters influencing the separation were studied, such as the concentrations of SDS, GMWNTs, and isopropanol; choice of carbon nanotubes; sodium borate content; and buffer pH. The results revealed that the presence of SC-GMWNTs in buffer enhanced the separation efficiency for the target analytes relative to conventional micelles due to the strong interaction between the surface of the GMWNTs and the target compounds. Under the optimum conditions, the method showed good linearity, with correlation coefficients higher than 0.9950. LODs were in the range of 0.71-3.10 μg/mL. Furthermore, satisfactory separations were achieved with good recovery values in the range of 89.97 and 103.30% when 10 mM borate, 30 mM SDS, 10% isopropanol, and 6 μg/mL SC-GMWNTs were introduced into the buffer solution.
Product Description
Supelco
Salvianolic acid A, analytical standard
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Protocatechuic acid, primary reference standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Salvianolic acid B, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%