Merck
  • Malva sylvestris L. extract suppresses desferrioxamine-induced PGE₂ and PGD₂ release in differentiated U937 cells: the development and validation of an LC-MS/MS method for prostaglandin quantification.
Malva sylvestris L. extract suppresses desferrioxamine-induced PGE₂ and PGD₂ release in differentiated U937 cells: the development and validation of an LC-MS/MS method for prostaglandin quantification.
Biomedical chromatography : BMC (2014-01-10)
Cleverson Antonio Ferreira Martins, Almeriane Maria Weffort-Santos, João Cleverson Gasparetto, Angela Cristina Leal Badaró Trindade, Michel Fleith Otuki, Roberto Pontarolo
ABSTRACT
Malva sylvestris is a species used worldwide as an alternative to anti-inflammatory therapies; however, its mechanism of action remains unknown. In this paper, the anti-inflammatory effects of M. sylvestris alcoholic extracts were evaluated by measuring the pro-inflammatory mediators PGE₂ and PGD₂ in desferrioxamine-stimulated phorbol 12-myristate 13-acetate-differentiated U937 cells. An HPLC-DAD fingerprint of the M. sylvestris extract was performed and caffeic acid, ferulic acid and scopoletin were identified and quantified. An HPLC-MS/MS method was developed and validated to separate and measure the prostaglandins. The lower limits of detection (~0.5 ng/mL for PGE₂ and PGD₂) and quantification (1.0 ng/mL for PGE₂ and PGD₂) indicated that the method is highly sensitive. The calibration curves showed excellent coefficients of correlation (r > 0.99) over the range of 1.0-500.0 ng/mL, and at different levels, the accuracy ranged from 96.4 to 106.4% with an RSD < 10.0% for the precision study. This method was successfully applied using U937-d cells. A significant dose-dependent reduction of PGE2 and PGD2 levels occurred using 10 µg/mL (10.74 ± 2.86 and 9.60 ± 6.89%) and 50 µg/mL of extract (48.37 ± 3.24 and 53.06 ± 6.15%), suggesting that the anti-inflammatory mechanisms evoked by M. sylvestris may be related to modulation of these mediators.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Prostaglandin D2, ≥95%, synthetic
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications