Merck
  • Crocin prevents sesamol-induced oxidative stress and apoptosis in human platelets.
Crocin prevents sesamol-induced oxidative stress and apoptosis in human platelets.
Journal of thrombosis and thrombolysis (2014-04-08)
Ram M Thushara, Mahadevappa Hemshekhar, Manoj Paul, Mahalingam Shanmuga Sundaram, Rohith L Shankar, Kempaiah Kemparaju, Kesturu S Girish
ABSTRACT
Recent studies have reported the platelet proapoptotic propensity of plant-derived molecules such as, resveratrol, thymoquinone, andrographolide and gossypol. Meanwhile, there were also reports of phytochemicals such as cinnamtannin B1, which shows antiapoptotic effect towards platelets. Platelets are mainly involved in hemostasis, thrombosis and wound healing. However, altered platelet functions can have serious pathological outcomes that include cardiovascular diseases. Platelets are sensitive to external and internal stimuli including therapeutic and dietary components. The anuclear platelets do undergo apoptosis via mitochondrial pathway. However, exaggerated rate of platelet apoptosis could lead to thrombocytopenia and other bleeding disorders. The present study deals with ameliorative efficacy of crocin on sesamol-induced platelet apoptosis. The antiapoptotic property of crocin and the proapoptotic tendency of sesamol in platelets were previously demonstrated. Therefore, it was interesting to see how these two compounds would interact and wield their effects on human platelets. Crocin effectively inhibited sesamol-induced oxidative stress on platelets, which was evidenced by the measurement of endogenously generated reactive oxygen species, particularly hydrogen peroxide, and changes in thiol levels. Further, crocin abrogated sesamol-induced biochemical events of apoptosis in platelets, which include intracellular calcium mobilization, changes in mitochondrial membrane integrity, cytochrome c release, caspase activity and phosphatidylserine externalization. Even though sesamol has proapoptotic effects on platelets, its anti-platelet activity cannot be neglected. Thus, the study proposes that sesamol could be supplemented with crocin, an approach that could not only abolish the toxic effects of sesamol on platelets, but also enhance the quality of treatment due to their synergistic action.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Benzamidine hydrochloride, 99%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Crocin, suitable for microscopy
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Rotenone, ≥95%
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)