Merck
Six new alkaloids from Melodinus henryi.
Fitoterapia (2014-12-08)
Ke Ma, Jun-Song Wang, Jun Luo, Ling-Yi Kong
ABSTRACT
A total of six new alkaloids, melodinhenines A-F (1-6), were isolated from Melodinus henryi. Melodinhenines A and B are new eburnan-vindolinine-type bisindole alkaloids and melodinhenines C-F are new quinolinic melodinus alkaloids. Their structures were elucidated through extensive spectroscopic methods including 2D NMR and HRESIMS analyses. The absolute configuration of 1 and 2 was determined using ECD exciton chirality method. To the best of our knowledge, this is the first report on the determination of the absolute configuration of eburnan-vindolinine-type bisindole alkaloid using this method.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%