Merck
  • Upregulation of ANGPTL4 messenger RNA and protein in severely calcified carotid plaques.
Upregulation of ANGPTL4 messenger RNA and protein in severely calcified carotid plaques.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association (2013-10-01)
Hiroyuki Katano, Kazuo Yamada
ABSTRACT
In carotid atherosclerotic lesions, calcified plaques are thought to be stable and to evoke very few symptoms. However, the molecular activity in calcified plaques and their clinical significance have not been fully clarified yet. Carotid plaques from 18 endarterectomy patients were classified into high- and low-calcified plaques on the basis of Agatston calcium score. Twelve plaques were investigated for the alteration of gene expression by microarray analysis and real-time polymerase chain reaction (PCR) and 6 other plaques underwent protein assessment to elucidate the difference in molecular biological activity between the groups. Microarray analysis demonstrated 93 angiogenesis or growth factor-related transcripts that are reliably expressed (175 probe sets). Among them, angiopoietin-like protein 4 (ANGPTL4) expression was significantly elevated, whereas fibroblast growth factor receptor 2 (FGFR2) expression was significantly suppressed. Quantitative messenger RNA analysis was performed with real-time PCR. Augmented or decreased protein expression of each gene was confirmed by Western blotting analysis and immunohistochemistry. In high-calcified plaques, ANGPTL4 might be upregulated for antiangiogenic modulating function together with the downregulation of FGFR2, contributing to the stability of the plaques.
Product Description
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%