Merck
  • Suitable in vitro Eimeria arloingi macromeront formation in host endothelial cells and modulation of adhesion molecule, cytokine and chemokine gene transcription.
Suitable in vitro Eimeria arloingi macromeront formation in host endothelial cells and modulation of adhesion molecule, cytokine and chemokine gene transcription.
Parasitology research (2014-10-24)
Liliana M R Silva, Maria J M Vila-Viçosa, Helder C E Cortes, Anja Taubert, Carlos Hermosilla
ABSTRACT
Eimeria arloingi infections can cause severe haemorrhagic enteritis in young goat kids, thereby leading to high economic losses in goat industry worldwide. We aimed to isolate a new E. arloingi strain and establish a suitable in vitro culture system for the first merogony. E. arloingi oocysts were collected from naturally infected goat kids in the province of Alentejo, Portugal. For the maintenance of E. arloingi (strain A), kids kept under strict parasite-free conditions were orally infected with 10(3) sporulated oocysts each. Further, a new excystation protocol was successfully established to obtain viable sporozoites for further in vitro development in primary bovine umbilical vein endothelial cells (BUVEC). Overall, E. arloingi first merogony was successfully accomplished in BUVEC leading to macromeront formation (up to 150 μm) and the release of fully developed merozoites I stages. Moreover, host endothelial cell-parasite interactions were investigated in order to determine the extent of modulation carried out by E. arloingi in BUVEC during the first merogony. Gene transcription of adhesion molecules (E-selectin, P-selectin, VCAM-1, ICAM-1) was enhanced in the first hours post-infection (p.i.) in E. arloingi-infected BUVEC. BUVEC activation due to invasion was also shown by increased chemokine (CXCL8, CCL2, CCL5), cytokine (GM-CSF) and COX-2 gene transcription. The new E. arloingi (strain A) will be useful for better comprehension of early host innate immune reactions against this parasite in vitro/in vivo as well as to further our investigations in the complex Eimeria-host endothelial cell interactions.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Oleic acid, analytical standard
Oleic acid, European Pharmacopoeia (EP) Reference Standard