Merck
  • D-serine-induced inactivation of NMDA receptors in cultured rat hippocampal neurons expressing NR2A subunits is Ca2+-dependent.
D-serine-induced inactivation of NMDA receptors in cultured rat hippocampal neurons expressing NR2A subunits is Ca2+-dependent.
CNS neuroscience & therapeutics (2014-07-22)
Xia Li, Yuan-Yuan Zhang, Zhao-Qin Chen, Zheng-Lin Jiang, Li Sun, Li-Hua Xu, Yao Yang, Yun-Feng Zhang
ABSTRACT
Our previous studies indicate that glycine can inhibit N-methyl-D-aspartate receptor (NMDAR) responses induced by high concentrations of NMDA in rat hippocampal neurons. The present study was designed to observe whether D-serine induces inactivation of NMDARs in cultured rat hippocampal neurons and to investigate the underlying mechanisms of this effect. Cell culture, whole-cell patch-clamp electrophysiology, Ca(2+) imaging, immunohistochemistry, and Western blot analysis were used. We found that the peak current and Ca(2+) influx evoked by 30 μM NMDA were increased by co-application of D-serine, but those evoked by 300 μM NMDA were reduced dose-dependently by co-application of D-serine. However, the inhibitory effect of D-serine on NMDAR responses was reversed by ZnCl2 (30 nM), an inhibitor of the NR2A subunit, but was less influenced by ifenprodil (10 μM), an NR2B inhibitor. In addition, the inhibitory effect of D-serine was not detected in young hippocampal neurons that expressed less of the NR2A subunits and reversed in the presence of 10 mM BAPTA. D-serine can also induce inactivation of NMDARs, the NR2A subunit is required for the induction of this effect, and this inactivation is Ca(2+)-dependent in nature. This action of D-serine is hypothesized to play a neuroprotective role upon a sustained large glutamate insult to the brain.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%