Merck
  • Depression and anxiety in women with epilepsy during pregnancy and after delivery: a prospective population-based cohort study on frequency, risk factors, medication, and prognosis.
Depression and anxiety in women with epilepsy during pregnancy and after delivery: a prospective population-based cohort study on frequency, risk factors, medication, and prognosis.
Epilepsia (2014-12-20)
Marte Helene Bjørk, Gyri Veiby, Simone C Reiter, Jan Øystein Berle, Anne Kjersti Daltveit, Olav Spigset, Bernt A Engelsen, Nils Erik Gilhus
ABSTRACT
To assess incidence, prevalence, risk factors, and prognosis of peripartum depression and anxiety in a prospective study of women with epilepsy. Pregnancies in women with epilepsy (n=706) were compared to pregnancies in all women without epilepsy (n=106,511) including women with specified nonepileptic chronic diseases (n=8,372) in the Norwegian Mother and Child Cohort Study. The database was linked to the Medical Birth Registry of Norway. Depression and anxiety were assessed with validated questionnaires five times from the second trimester to 36 months after delivery. Blood was drawn for analysis of antiepileptic drug (AED) concentrations. Women with epilepsy more often had peripartum depression (26.7%) or anxiety (22.4%) than women without epilepsy (18.9% and 14.8%, respectively, p<0.001 for both comparisons) and women with other chronic diseases (23.1% and 18.4%, respectively, p=0.03 and 0.01). Women using AEDs during pregnancy were especially at risk regardless of AED type. The risk further increased with the use of multiple AEDs and with high doses and/or plasma levels. Risk factors associated with peripartum depression and/or anxiety in the epilepsy cohort were high seizure frequency, a history of physical and/or sexual abuse, adverse socioeconomic factors, previous loss of a child, AED use, unplanned pregnancy, and prepregnancy depression and/or anxiety. The recovery rate 3 years after delivery was lower for women with epilepsy with a history of depression/anxiety or physical/sexual abuse than for women without epilepsy. Depressed women with epilepsy were less frequently treated with antidepressive drugs during pregnancy than women without epilepsy. Women with epilepsy frequently have depression and anxiety during and after pregnancy. Patients at risk should be identified before delivery as depressive symptoms could be undertreated in this group.
Product Description
Levetiracetam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Levetiracetam, ≥98% (HPLC)
Supelco
Lamotrigine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Lamotrigine for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Lamotrigine, United States Pharmacopeia (USP) Reference Standard
Lamotrigine for system suitability, European Pharmacopoeia (EP) Reference Standard
Lamotrigine, European Pharmacopoeia (EP) Reference Standard
USP
Levetiracetam, United States Pharmacopeia (USP) Reference Standard
Supelco
Levetiracetam, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Oxcarbazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Oxcarbazepine, ≥98% (HPLC), solid
Sigma-Aldrich
Carbamazepine, powder
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Oxcarbazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard