Merck
  • Toxicological assessment of kretek cigarettes: Part 1: background, assessment approach, and summary of findings.
Toxicological assessment of kretek cigarettes: Part 1: background, assessment approach, and summary of findings.
Regulatory toxicology and pharmacology : RTP (2014-12-17)
E Roemer, R Dempsey, M K Schorp
ABSTRACT
This publication introduces a series of six other publications describing the toxicological assessment of kretek cigarettes, i.e., cigarettes characterized primarily by the use of a significant amount of cloves as an ingredient added to the tobacco. This paper presents background information on kretek cigarettes, describes the general approach of the in vitro and in vivo toxicological assessment of mainstream smoke from kretek cigarettes, presents the methodology used, and summarizes the results of the assessment program. In summary, the smoke from kretek cigarettes gives rise to the typical cigarette smoke-related effects known from American-blended cigarettes, does not reveal any novel toxicity, and exhibits an unexpected distinct attenuation of pulmonary inflammation. Based on equal amounts of smoke total particulate matter (TPM), kretek cigarettes deliver less toxicants when compared to American-blended cigarettes; when assessed in vitro, the smoke from kretek cigarettes is less cytotoxic (gas/vapor phase) and less mutagenic (TPM). When assessed in vivo, kretek cigarette smoke shows lower toxicity in the respiratory tract. When based on an equal nicotine basis, several of the toxicity endpoints in kretek cigarettes become equivalent to American-blended cigarettes. The data do not indicate an increased hazard potential of kreteks compared to American-blended cigarettes.
Sigma-Aldrich
Acetamide, sublimed, 99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
o-Anisidine, ≥99%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Rhein, technical grade
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Acetamide, ≥99.0% (GC)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Supelco
Styrene, analytical standard
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
SAFC
Isoprene, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Isoprene, 99%, contains <1000 ppm p-tert-butylcatechol as inhibitor
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder