Merck
  • Comparison of xenobiotic metabolizing enzyme activities in ex vivo human skin and reconstructed human skin models from SkinEthic.
Comparison of xenobiotic metabolizing enzyme activities in ex vivo human skin and reconstructed human skin models from SkinEthic.
Archives of toxicology (2014-03-25)
Joan Eilstein, Guillaume Léreaux, Natali Budimir, Georges Hussler, Simon Wilkinson, Daniel Duché
ABSTRACT
Skin function is not limited to a physical barrier. According to its total surface area, it is also considered as an extra-hepatic metabolizing organ. In vitro engineered human skins have been developed to replace limited ex vivo normal human skin samples (NHS). Thus, assessing and comparing skin models from SkinEthic [Episkin™, RHE™ and the full thickness model (FTM)] with NHS in terms of metabolic capability are essential. The apparent activities of main cutaneous isoforms of cytochrome P450-dependent monooxygenases (CYP1A1/1B1, 2B6/2C18/2E1, 3A5/3A7), esterase, glutathione-S-[(GST), A, M, P, T], N-acetyl-(NAT1), uridinyl-diphosphate glucuronyl-(UDPGT 1A family) and sulfo-(SULT1A1) transferases were determined using probe substrates. Mean activities indicative of CYP1A1/1B1 (expressed as pmol/mg protein/6 h) in RHE™ (2.8) and FTM (2.6) were very similar to NHS (3.0) while Episkin™ showed a higher activity (9.1). Activities of CYP3A5/3A7 in FTM (3.3) and Episkin™ (3.6) were similar to NHS (3.8) while activity in RHE™ (13.3) was higher. CYP2B6/2C18/2E1 activity was below LOQ (0.5) in all skin models and NHS. Comparable intrinsic metabolic clearances were measured between NHS and skin models for esterase, UDPGT, GST and NAT1 activities. SULT1A1 activity toward probe substrates was not detected in skin models and observed at the limit of detection in NHS. Weak cytochrome P450-dependent monooxygenases, high esterase and transferase activities suggested that NHS and skin models exhibited limited functionalization and much greater detoxification (hydrolytic and conjugating) capacities. These results demonstrate that skin models are similar to NHS in terms of metabolic functionality toward xenobiotics investigated and useful tools to assess both the local efficiency and safety of cosmetics.
Product Description
Coumarin, primary reference standard
Sigma-Aldrich
Allyl sulfide, ≥97%, FG
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
4-Acetamidobenzoic acid, 98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Allyl sulfide, 97%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
4-Aminobenzoic acid sodium salt, ≥99%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Coumarin, ≥99% (HPLC)