Merck
  • Aberrant liver insulin receptor isoform a expression normalises with remission of type 2 diabetes after gastric bypass surgery.
Aberrant liver insulin receptor isoform a expression normalises with remission of type 2 diabetes after gastric bypass surgery.
PloS one (2015-03-06)
Vinko Besic, Hongjun Shi, Richard S Stubbs, Mark T Hayes
ABSTRACT
Type 2 diabetes mellitus (T2DM) results from a combination of progressive insulin resistance and loss of pancreatic beta cell function and/or mass. Insulin signalling occurs through the insulin receptor, (INSR) which is alternatively spliced into two isoforms: INSRA (-exon 11) and INSRB (+exon 11). Because the INSR isoforms have different functional characteristics, their relative expression ratio has been implicated in the pathogenesis of insulin resistance and T2DM. We studied levels of INSR isoform mRNA in liver samples taken from 46 individuals with or without T2DM at Roux-en-Y (RYGB) surgery, and on average 17 (± 5.6) months later in 16 of the same individuals (8 diabetic and non-diabetic patients). INSRA or INSRB was also overexpressed in HepG2 cells to ascertain their effect on AKT phosphorylation and PCK1 expression as markers of insulin-mediated metabolic signalling. We found the INSRB:A isoform ratio was reduced in individuals with T2DM in comparison to those with normal glucose tolerance and normalised with remission of diabetes. The INSRB:A ratio increased due to a reduction in the alternatively spliced INSRA isoform following remission of diabetes. Overexpressing INSRA isoform in HepG2 hepatoma cells reduced inhibition of PCK1 transcription and did not increase AKT phosphorylation in response to insulin load compared to the effect of overexpressing the B isoform. Data presented here revitalizes the role of the INSR isoforms in the pathogenesis of T2DM, and suggests that an abrogated INSRB:A ratio that favours the INSRA isoform may negatively impact insulin-mediated metabolic signalling.
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)