Merck
  • ATL-1, a synthetic analog of lipoxin, modulates endothelial permeability and interaction with tumor cells through a VEGF-dependent mechanism.
ATL-1, a synthetic analog of lipoxin, modulates endothelial permeability and interaction with tumor cells through a VEGF-dependent mechanism.
Biochemical pharmacology (2014-06-03)
A M Vieira, E H Neto, C C Figueiredo, C Barja Fidalgo, I M Fierro, V Morandi
ABSTRACT
Lipoxins (LX) and 15-epi-LX are lipids with a potent inhibitory effect on angiogenesis, in different models in vivo and in vitro. ATL-1, a synthetic analog of 15-epi-LXA4, inhibits various actions stimulated by vascular endothelial growth factor (VEGF). However, LX actions on endothelial cells (EC) in tumor-related contexts are still unknown. Here, we investigated the modulation of EC by ATL-1, in a model that mimics tumor extravasation. We observed that the analog inhibited endothelial permeability induced by VEGF, through the stabilization of VE-cadherin/β-catenin-dependent adherens junctions. We tested the ability of MV3 cells, a highly metastatic melanoma cell line, to transmigrate across unchallenged EC monolayers for 18 h, as compared to NGM normal melanocytes. ATL-1 was able to inhibit only melanoma extravasation. MV3 cells secrete large amounts of VEGF and we observed that ATL-1 per se did not alter this ability. Melanoma cells skills to crossing endothelial monolayers were due to the steady accumulation of tumor-derived VEGF. When endothelial cells were challenged with exogenous VEGF, added at levels comparable to those secreted by MV3 cells over 18 h, and a short-term (4h) transendothelial migration assay was performed, both melanoma and melanocyte cells were able to extravasate, and ATL-1 was able to block the passage of both cells. These results indicate that ATL-1 has a potent inhibitory effect on the permeability induced by VEGF, and that this pharmacological effect could be used to block tumor extravasation across endothelial barriers, with a possible prospect of reducing the haematogenic spread of cancer cells.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O