Merck
  • Solid lipid nanoparticles-loaded topical gel containing combination drugs: an approach to offset psoriasis.
Solid lipid nanoparticles-loaded topical gel containing combination drugs: an approach to offset psoriasis.
Expert opinion on drug delivery (2014-08-01)
Rahul Sonawane, Harshad Harde, Mahesh Katariya, Satyam Agrawal, Sanyog Jain
ABSTRACT
The primary aim of present work was to develop effective combination drug therapy for topical treatment of psoriasis. Betamethasone dipropionate and calcipotriol loaded solid lipid nanoparticles (CT-BD-SLNs) were prepared by hot melt high shear homogenization technique, which were then incorporated in Carbopol gel matrix. The anti-psoriatic potential was tested by sequential in vitro (skin permeation and dermal distribution, anti-proliferative effect in HaCaT cells) and in vivo (Draize patch irritation, transepidermal water loss (TEWL) and anti-psoriatic mouse tail studies) experiments. A negligible amount in receptor compartment, yet confined distribution of drugs to epidermal and dermal region of skin was observed in case of SLNs, which is essential for safe and effective anti-psoriatic therapy. Draize patch test and TEWL demonstrated negligible skin irritation and better skin tolerability of SLNs. The in vitro HaCaT cell line study demonstrated that SLNs delayed the abrupt growth of keratinocytes, while in vivo mouse tail model showed that SLNs gel significantly decreased the epidermal thickness and increased melanocyte count in comparison to commercial Daivobet® ointment. The developed SLNs gel is expected to be potential strategies for treatment of psoriasis and other topical diseases.
Product Description
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium metabisulfite, analytical standard
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Trolamine, European Pharmacopoeia (EP) Reference Standard
Tristearin, European Pharmacopoeia (EP) Reference Standard
USP
Sodium metabisulfite, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%