Merck
  • The unimolecular chemistry of protonated and deprotonated 2,2-dinitroethene-1,1-diamine (FOX-7) studied by tandem mass spectrometry and computational chemistry.
The unimolecular chemistry of protonated and deprotonated 2,2-dinitroethene-1,1-diamine (FOX-7) studied by tandem mass spectrometry and computational chemistry.
European journal of mass spectrometry (Chichester, England) (2014-06-04)
Ján Žabka, Ludmila Šimková, Zdeněk Jalový, Miroslav Polášek
ABSTRACT
2,2-Dinitroethene-1,1-diamine (FOX-7) was studied by means of electrospray ionization (ESI) and chemical ionization (CI) mass spectrometry in both positive and negative ion mode. Detailed mechanisms of unimolecular fragmentations of protonated and deprotonated FOX-7 were investigated using high- and low- energy collision-induced dissociation (CID) mass spectrometry, neutral fragment reionization mass spectrometry and quantum chemistry calculations. In deprotonated FOX-7, elimination of the carbodiimide molecule was identified as the energetically most favored fragmentation channel, closely resembling the base hydrolysis of FOX-7. The dinitromethanide ion is formed during this fragmentation as revealed by comparison with CID mass spectra of an isobaric ion prepared by the ESI of authentic sodium dinitromethanide. The proton affinity of FOX-7 was estimated as 855 kJ mo(-1) by high-accuracy quantum chemistry calculations. This value corresponds to protonation at the C-2 position, though the oxygen-protonated tautomer was found to be nearly isoenergetic in the gas phase. In acetonitrile, the nitro group-protonated FOX-7 was found to be significantly less stable then its C-2 tautomer. These theoretical findings are clearly reflected in differences in fragmentations of ESI- and CI-generated [M+H(]+) ions. Interestingly, the consecutive losses of OH∙ and NO2∙ radicals instead of a whole HNO3 molecule were found to account for the most abundant fragment ion in the positive ESI CID mass spectra. In the CI-generated [M+H](+) and [M+D](+) ions, substantial internal energy effects upon the CID were observed.
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Acetone-d6, "100%", 99.95 atom % D
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Acetone-d6, 99.5 atom % D
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Acetone-d6, 99.9 atom % D
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Acetone-d6, 99.9 atom % D
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.99 atom % 12C
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon