Merck
  • Similar survival, but better quality of life after myeloablative transplantation using unrelated cord blood vs matched sibling donors in adults with hematologic malignancies.
Similar survival, but better quality of life after myeloablative transplantation using unrelated cord blood vs matched sibling donors in adults with hematologic malignancies.
Bone marrow transplantation (2014-05-21)
H-L Liu, Z-M Sun, L-Q Geng, X-B Wang, K-Y Ding, J Tong, B-L Tang, C-Y Zhou, W Yao, C-C Zheng, K-D Song, X-Y Zhu, W Wei, Y Wang
ABSTRACT
This study included data from 185 consecutively treated patients, 16 years of age or older, who underwent myeloablative transplantation using unrelated umbilical cord blood (UCB) (UCB transplantation (UCBT), n=70) or HLA-identical sibling donor peripheral blood stem cells alone or combined with bone marrow (BMT/PBSCT, n=115) from October 2001 to December 2012. All patients received myeloablative regimens, cyclosporin A plus mycophenolate mofetil as prophylaxis for GVHD, and similar supportive care. Although hematopoietic recovery was significantly delayed after UCBT, the rate of neutrophil engraftment was comparable. The median follow-up was 53 months (range, 15-136 months) for BMT/peripheral blood SCT (PBSCT) recipients and 35 months (range, 10-123 months) for UCBT recipients. There were no significant differences in the cumulative incidence of grades III to IV acute GVHD, relapse rate, or 3-year probabilities of disease-free survival between patients receiving UCBT and those receiving BMT/PBSCT. However, the cumulative incidence of chronic and extensive chronic GVHD was lower in UCBT recipients. The rates of long-term survivors returning to school or work and off immunosuppressive therapy were significantly higher after UCBT, which indicated that long-term survivors who underwent UCBT had a higher quality of life.
Product Description
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
USP
Mycophenolate mofetil, United States Pharmacopeia (USP) Reference Standard
Supelco
Mycophenolate Mofetil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard