Merck
  • Descending effect on spinal nociception by amygdaloid glutamate varies with the submodality of noxious test stimulation.
Descending effect on spinal nociception by amygdaloid glutamate varies with the submodality of noxious test stimulation.
Neuroscience letters (2014-04-22)
Nora Bourbia, Boriss Sagalajev, Antti Pertovaara
ABSTRACT
Amygdala has an important role in the processing of primary emotions, such as fear. Additionally, amygdala is involved in processing and modulation of pain. While the amygdala, particularly its central nucleus (CeA), has been shown to contribute to pain control, the descending pain regulation by the CeA is still only partly characterized. Here heat and mechanical nociception was tested in both hind limbs of healthy rats with a chronic guide cannula for microinjection of glutamate into the CeA of the left or right hemisphere. The aim was to assess whether the descending pain regulatory effect by glutamate in the amygdala varies with the submodality or the body side of nociceptive testing, brain hemisphere or the amygdaloid glutamate receptor. Motor performance was assessed with the Rotarod test. Amygdaloid glutamate, independent of the treated hemisphere, produced a dose-related heat and mechanical antinociception that varied with the submodality of testing. Heat antinociception was short lasting (minutes), bilateral and not reversed by blocking the amygdaloid NMDA receptor with MK-801. In contrast, mechanical antinociception lasted longer (>20 min), was predominantly contralateral and reversed by blocking the amygdaloid NMDA receptor. At an antinociceptive dose, amygdaloid glutamate failed to influence motor performance. The results indicate that independent of the brain hemisphere, the spatial extent and duration of the descending antinociceptive effect induced by amygdaloid glutamate varies with the amygdaloid glutamate receptor and the submodality of pain.
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
L-Glutamic acid monosodium salt hydrate, analytical standard