Merck
  • Lanostanoids with acetylcholinesterase inhibitory activity from the mushroom Haddowia longipes.
Lanostanoids with acetylcholinesterase inhibitory activity from the mushroom Haddowia longipes.
Phytochemistry (2015-01-13)
Shuang-Shuang Zhang, Qing-Yun Ma, Sheng-Zhuo Huang, Hao-Fu Dai, Zhi-Kai Guo, Zhi-Fang Yu, You-Xing Zhao
ABSTRACT
Nine lanostanoids, together with nine known ones, were isolated from the ethyl acetate extract of the fruiting bodies of the mushroom Haddowia longipes. Their structures were elucidated as 11-oxo-ganoderiol D, lanosta-8-en-7,11-dioxo-3β-acetyloxy-24,25,26-trihydroxy, lanosta-8-en-7-oxo-3β-acetyloxy-11β,24,25,26-tetrahydroxy, lanosta-7,9(11)-dien-3β-acetyloxy-24,25,26-trihydroxy, lanosta-7,9(11)-dien-3β-acetyloxy-24,26-dihydroxy-25-methoxy, 11-oxo-lucidadiol, 11β-hydroxy-lucidadiol, lucidone H and lanosta-7,9(11),24E-trien-3β-acetyloxy-26,27-dihydroxy by analysing their 1D/2D NMR and MS spectra. In addition, bioassays of inhibitory activity against acetylcholinesterase (AChE) of all compounds showed that thirteen compounds possessed inhibitory activity against AChE with the percentage inhibition ranging from 10.3% to 42.1% when tested at 100 μM.
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)