Merck
  • Proton pump inhibitors inhibit methotrexate transport by renal basolateral organic anion transporter hOAT3.
Proton pump inhibitors inhibit methotrexate transport by renal basolateral organic anion transporter hOAT3.
Drug metabolism and disposition: the biological fate of chemicals (2014-09-23)
Rym Chioukh, Marie-Sophie Noel-Hudson, Sandy Ribes, Natalie Fournier, Laurent Becquemont, Celine Verstuyft
ABSTRACT
The coadministration of methotrexate (MTX) and proton pump inhibitors (PPIs) can result in a pharmacokinetic interaction that delays MTX elimination and subsequently increases the MTX blood concentrations. Human organic anion transporters (hOATs) are responsible for the renal tubular secretion of MTX and are thought to be involved in this drug interaction. The aim of this study was to evaluate the inhibitory potencies of PPIs on hOAT1 and hOAT3, which are the two isoforms of OATs predominantly expressed in kidney proximal tubules. Using stably transfected cell systems that express the uptake transporters human embryonic kidney (HEK)-hOAT1 and HEK-hOAT3, we analyzed the inhibitory potencies of omeprazole, lansoprazole, and pantoprazole on OAT-mediated [(3)H]estrone sulfate (ES), [(3)H]p-aminohippuric acid (PAH), and [(3)H]MTX uptake in vitro. hOAT3 is a high affinity transporter for MTX (Km = 21.17 ± 5.65 µM). Omeprazole, lansoprazole, and pantoprazole inhibited [(3)H]MTX uptake in HEK-hOAT3 cells with an IC50 of 6.8 ± 1.16, 1.14 ± 0.26, and 4.45 ± 1.62 µM, respectively, and inhibited the [(3)H]ES uptake in HEK-hOAT3 cells with an IC50 of 20.59 ± 4.07, 3.96 ± 0.96, and 7.89 ± 2.31 µM, respectively. Furthermore, omeprazole, lansoprazole, and pantoprazole exhibited inhibited PAH uptake on hOAT1 in a concentration-dependent manner (IC50 = 4.32 ± 1.26, 7.58 ± 1.06, and 63.21 ± 4.74 µM, respectively). These in vitro results suggest that PPIs inhibit [(3)H]MTX transport via hOAT3 inhibition, which most likely explains the drug-drug interactions between MTX and PPIs and should be considered for other OATs substrates.
Product Description
Estrone, European Pharmacopoeia (EP) Reference Standard
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
p-Aminohippuric acid, ≥99%
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Probenecid, European Pharmacopoeia (EP) Reference Standard
USP
Probenecid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Omeprazole, European Pharmacopoeia (EP) Reference Standard
USP
Omeprazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Omeprazole, solid
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzimidazole, 98%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture