Merck
  • Branched amphiphilic peptide capsules: cellular uptake and retention of encapsulated solutes.
Branched amphiphilic peptide capsules: cellular uptake and retention of encapsulated solutes.
Biochimica et biophysica acta (2014-02-26)
Pinakin Sukthankar, L Adriana Avila, Susan K Whitaker, Takeo Iwamoto, Alfred Morgenstern, Christos Apostolidis, Ke Liu, Robert P Hanzlik, Ekaterina Dadachova, John M Tomich
ABSTRACT
Branched amphiphilic peptide capsules (BAPCs) are peptide nano-spheres comprised of equimolar proportions of two branched peptide sequences bis(FLIVI)-K-KKKK and bis(FLIVIGSII)-K-KKKK that self-assemble to form bilayer delimited capsules. In two recent publications we described the lipid analogous characteristics of our BAPCs, examined their initial assembly, mode of fusion, solute encapsulation, and resizing and delineated their capability to be maintained at a specific size by storing them at 4°C. In this report we describe the stability, size limitations of encapsulation, cellular localization, retention and, bio-distribution of the BAPCs in vivo. The ability of our constructs to retain alpha particle emitting radionuclides without any apparent leakage and their persistence in the peri-nuclear region of the cell for extended periods of time, coupled with their ease of preparation and potential tune-ability, makes them attractive as biocompatible carriers for targeted cancer therapy using particle emitting radioisotopes. This article is part of a Special Issue entitled: Interfacially Active Peptides and Proteins. Guest Editors: William C. Wimley and Kalina Hristova.
Product Description
Supelco
Rhodamine B, analytical standard
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamine B, suitable for fluorescence
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
2,5-Dihydroxybenzoic acid, analytical standard