Merck
  • Hesperetin inhibits rat coronary constriction by inhibiting Ca(2+) influx and enhancing voltage-gated K(+) channel currents of the myocytes.
Hesperetin inhibits rat coronary constriction by inhibiting Ca(2+) influx and enhancing voltage-gated K(+) channel currents of the myocytes.
European journal of pharmacology (2014-04-23)
Yu Liu, Longgang Niu, Lijuan Cui, Xiaomin Hou, Jiangtao Li, Xuanping Zhang, Mingsheng Zhang
ABSTRACT
Hesperetin (HSP, one of the most common flavonoids in Citrus) has been reported to possess many benificial effects and is indicated for many diseases both as a therapeutic drug and as a supplement. Although its vascular effects have been extensively studied, little is known about its effects and the underlying mechanisms on coronary artery. In the present study, the myogenic effects of HSP were studied with a wire myograph in isolated rat coronary artery (RCA). Molecular probe and the patch clamp technique were used to study effects of HSP on intracellular free Ca(2+) concentration, inward Ca(2+) currents through L-type voltage-gated Ca(2+) channels (LVGC) and outward K(+) currents through voltage-gated K(+) channels (KV). HSP (0.01-0.1mM) concentration-dependently depressed concentration-contraction curves of both KCl and thromboxane receptor agonist 9,11-Dideoxy-9α,11α-methanoepoxy prostaglandin F2α (U46619), and relaxed RCA precontracted by the both vasoconstrictors. The vasospasmolytic effect was more potent in KCl- than in U46619-induced contraction. The vasorelaxation was attenuated by 4-aminopyridine, a specific KV inhibitor, but not affected by NG-nitro-L-arginine methylester ester, indomethacin, glibenclamide, iberiotoxin, BaCl2 or endothelium denudation. At the same concentrations, HSP inhibited extracellular Ca(2+) influx-induced contraction, reduced intracellular free Ca(2+) concentration, inhibited inward Ca(2+) currents through LVGC and increased outward K(+) currents through KV in the vascular smooth muscle cells (VSMCs) freshly isolated from RCA. Collectively, our results show that HSP is vasospasmolytic in RCA and suggest that the vasospasmolysis is mediated by inhibition of LVGC and enhancement of KV currents in RCA VSMCs.
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material