Merck
  • Biochemical and pharmacological characterization of three toxic phospholipase A2s from Daboia russelii snake venom.
Biochemical and pharmacological characterization of three toxic phospholipase A2s from Daboia russelii snake venom.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2014-12-06)
J R Kumar, Balapal S Basavarajappa, B S Vishwanath, T Veerabasappa Gowda
ABSTRACT
Three isoenzymes of phospholipase A2 (PLA2), VRV-PL-IIIc, VRV-PL-VII, and VRV-PL-IX were isolated from Daboia russelii snake venom. The venom, upon gel filtration on Sephadex G-75 column, resolved into six peaks (DRG75 I-VI). The VRV-PL-IIIc was purified by subjecting DRG75II to homogeneity by rechromatography in the presence of 8M urea on Sephadex G-75 column. The other two isoenzymes VRV-PL-VII and VRV-PL-IX were purified by subjecting DRG75III to ion exchange chromatography on CM-Sephadex C-25 column. Mol wt. for the three PLA2s, VRV-PL-IIIc, VRV-PL-VII, and VRV-PL-IX are 13.003kDa, 13.100kDa and 12.531kDa respectively. The VRV-PL-IIIc is not lethal to mice up to 14mg/kg body weight but it affects blood sinusoids and causes necrosis of the hepatocytes in liver. It causes hemorrhage in kidney and shrinkage of renal corpuscles and renal tubules. The LD50s for VRV-PL-VII and VRV-PL-IX are 7 and 7.5mg/kg body weight respectively. They induced neurotoxic symptoms similar to VRV-PL-V. All the three PLA2s are anticoagulant and induced varying degree of edema in the foot pads of mice. VRV-PL-V and VRV-PL-VII are shown to act as pre and post synaptic toxins, while VRV-PL-IX acts as presynaptic toxin. This is evident from experiments conducted on cultured hippocampal neurons by patch clamp electrophysiology.
Product Description
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)