Merck
  • Quantification of cocaine and metabolites in exhaled breath by liquid chromatography-high-resolution mass spectrometry following controlled administration of intravenous cocaine.
Quantification of cocaine and metabolites in exhaled breath by liquid chromatography-high-resolution mass spectrometry following controlled administration of intravenous cocaine.
Analytical and bioanalytical chemistry (2014-08-19)
Kayla N Ellefsen, Marta Concheiro, Olof Beck, David A Gorelick, Sandrine Pirard, Marilyn A Huestis
ABSTRACT
Breath has been investigated as an alternative matrix for detecting recent cocaine intake; however, there are no controlled cocaine administration studies that investigated the drug's disposition into breath. Breath was collected from 10 healthy adult cocaine users by asking them to breathe into a SensAbues device for 3 min before and up to 22 h following 25 mg intravenous (IV) cocaine dosing on days 1, 5, and 10, and assayed with a validated liquid chromatography-high-resolution mass spectrometry (LC-HRMS) method to quantify breath cocaine, benzoylecgonine (BE), ecgonine methyl ester (EME), and norcocaine. The assay was linear from 25 to 1,000 pg/filter, extraction efficiencies were 83.6-126%, intra- and inter-assay imprecision was <10.6%, and bias was between -8.5 and 16.8%. No endogenous or exogenous interferences were observed for more than 75 tested. Analytes were generally stable under short-term storage conditions. Ion suppression was less than 46%. Of breath specimens collected after controlled cocaine administration, 2.6% were positive for cocaine (26.1-66 pg/filter, 1-9.5 h), 0.72% BE (83.3-151 pg/filter, 6.5-12.5 h), and 0.72% EME (50-69.1 pg/filter, 6.5-12.5 h); norcocaine was not detected. Methanolic extraction of the devices themselves, after filters were removed, yielded 19.2% positive cocaine tests (25.2-36.4 pg/device, 10 min-22 h) and 4.3% positive BE tests (26.4-93.7 pg/device, 10 min-22 h), explaining differences between the two extraction techniques. These results suggest that the device reflects the drug in oral fluid as well as lung microparticles, while the filter reflects only drug-laden microparticles. A sensitive and specific method for cocaine, BE, EME, and norcocaine quantification in breath was developed and validated. Cocaine in breath identifies recent cocaine ingestion, but its absence does not preclude recent use.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard