Merck
  • Hydrastine pharmacokinetics and metabolism after a single oral dose of goldenseal (Hydrastis canadensis) to humans.
Hydrastine pharmacokinetics and metabolism after a single oral dose of goldenseal (Hydrastis canadensis) to humans.
Drug metabolism and disposition: the biological fate of chemicals (2015-01-23)
Prem K Gupta, Gary Barone, Bill J Gurley, E Kim Fifer, Howard P Hendrickson
ABSTRACT
The disposition and metabolism of hydrastine was investigated in 11 healthy subjects following an oral dose of 2.7 g of goldenseal supplement containing 78 mg of hydrastine. Serial blood samples were collected for 48 hours, and urine was collected for 24 hours. Hydrastine serum and urine concentrations were determined by Liquid Chromatography-tandem mass spectrometry (LC-MS/MS). Pharmacokinetic parameters for hydrastine were calculated using noncompartmental methods. The maximal serum concentration (Cmax) was 225 ± 100 ng/ml, Tmax was 1.5 ± 0.3 hours, and area under the curve was 6.4 ± 4.1 ng ⋅ h/ml ⋅ kg. The elimination half-life was 4.8 ± 1.4 hours. Metabolites of hydrastine were identified in serum and urine by using liquid chromatography coupled to high-resolution mass spectrometry. Hydrastine metabolites were identified by various mass spectrometric techniques, such as accurate mass measurement, neutral loss scanning, and product ion scanning using Quadrupole-Time of Flight (Q-ToF) and triple quadrupole instruments. The identity of phase II metabolites was further confirmed by hydrolysis of glucuronide and sulfate conjugates using bovine β-glucuronidase and a Helix pomatia sulfatase/glucuronidase enzyme preparation. Hydrastine was found to undergo rapid and extensive phase I and phase II metabolism. Reduction, O-demethylation, N-demethylation, hydroxylation, aromatization, lactone hydrolysis, and dehydrogenation of the alcohol group formed by lactone hydrolysis to the ketone group were observed during phase I biotransformation of hydrastine. Phase II metabolites were primarily glucuronide and sulfate conjugates. Hydrastine undergoes extensive biotransformation, and some metabolites may have pharmacological activity. Further study is needed in this area.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium formate, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Isoquinoline, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Methanol, Absolute - Acetone free