Merck
  • The glycoprotein Ten-m3 mediates topography and patterning of thalamostriatal projections from the parafascicular nucleus in mice.
The glycoprotein Ten-m3 mediates topography and patterning of thalamostriatal projections from the parafascicular nucleus in mice.
The European journal of neuroscience (2014-11-19)
Heidi Tran, Atomu Sawatari, Catherine A Leamey
ABSTRACT
The striatum is the key input nucleus of the basal ganglia, and is implicated in motor control and learning. Despite the importance of striatal circuits, the mechanisms associated with their development are not well established. Previously, Ten-m3, a member of the Ten-m/teneurin/odz family of transmembrane glycoproteins, was found to be important in the mapping of binocular visual pathways. Here, we investigated a potential role for Ten-m3 in striatal circuit formation. In situ hybridisation revealed a patchy distribution of Ten-m3 mRNA expression superimposed on a high-dorsal to low-ventral gradient in a subregion of the striatal matrix. A survey of afferent/efferent structures associated with the matrix identified the parafascicular thalamic nucleus (PF) as a potential locus of action. Ten-m3 was also found to be expressed in a high-dorsal to low-ventral gradient in the PF, corresponding topographically to its expression in the striatum. Further, a subset of thalamic terminal clusters overlapped with Ten-m3-positive domains within the striatal matrix. Studies in wild-type (WT) and Ten-m3 knockout (KO) mice revealed no differences in overall striatal or PF structure. Thalamostriatal terminals in KOs, however, while still confined to the matrix subregion, lost their clustered appearance. Topography was also altered, with terminals from the lateral PF projecting ectopically to ventral and medial striatum, rather than remaining confined dorsolaterally as in WTs. Behaviorally, Ten-m3 KOs displayed delayed motor skill acquisition. This study demonstrates that Ten-m3 plays a key role in directing the formation of thalamostriatal circuitry, the first molecular candidate reported to regulate connectivity within this pathway.
Product Description
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Fluorescein, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard