Merck
  • Total monoamine oxidase (MAO) inhibition by chestnut honey, pollen and propolis.
Total monoamine oxidase (MAO) inhibition by chestnut honey, pollen and propolis.
Journal of enzyme inhibition and medicinal chemistry (2013-10-26)
O Yildiz, F Karahalil, Z Can, H Sahin, S Kolayli
ABSTRACT
Monoamine oxidase (MAO) inhibitors are generally used in the treatment of depressive disorders and some neurodegenerative illnesses, such as Parkinson's disease and Alzheimer's disease. The aim of this preliminary study was to investigate the MAO [MAO (E.C.1.4.3.4)] inhibiting effect of various apitherapeutic products, such as chestnut honey, pollen and propolis. Extracts' MAO inhibition was measured using peroxidase-linked spectrophotometric assay in enzyme isolated from rat liver microsomes, and the values are expressed as the inhibition concentration (IC50) causing 50% inhibition of MAO. The antioxidant activity of the bee products was also determined in terms of total phenolic content (TPC) and ferric reducing/antioxidant power in aquatic extracts. All samples exhibited substantial inhibition of MAO, propolis having the highest. Inhibition was related to samples' TPCs and antioxidant capacities. These results show that bee products possess a sedative effect and may be effective in protecting humans against depression and similar diseases.
Product Description
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Supelco
4-Aminoantipyrine, analytical standard
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG