Merck
  • Low doses of dextromethorphan have a beneficial effect in the treatment of neuropathic pain.
Low doses of dextromethorphan have a beneficial effect in the treatment of neuropathic pain.
Fundamental & clinical pharmacology (2014-04-08)
Véronique Morel, Gisèle Pickering, Monique Etienne, Amandine Dupuis, Anne-Marie Privat, Maryse Chalus, Alain Eschalier, Laurence Daulhac
ABSTRACT
N-methyl-D-aspartate receptor (NMDAR) antagonists may be given in persistent neuropathic pain, but adverse events especially with ketamine may limit their clinical use. Less central and cognitive adverse events are described with dextromethorphan and memantine. These molecules have been explored in many preclinical and clinical studies, but data are conflicting as regards neuropathic pain alleviation. Dextromethorphan and memantine have been administered to animals after spinal nerve ligation (SNL) to evaluate their antinociceptive/cognitive effects and associated molecular events, including the phosphorylation of several tyrosine (pTyr(1336), pTyr(1472)) residues in the NR2B NMDAR subunit. Spinal nerve ligation and sham animals received dextromethorphan (10 mg/kg, i.p.), memantine (20 mg/kg, i.p.) or saline (1 mL/kg, i.p.). These drugs were administered once symptoms of allodynia and hyperalgesia had developed. Tests were carried out before and after surgery. Tactile allodynia, mechanical hyperalgesia and spatial memory were, respectively, evaluated by von Frey, Randall & Selitto and Y-maze tests and molecular events by Western blot analysis. Spinal nerve-ligated animals displayed nociception and impaired spatial memory. Dextromethorphan, but not memantine, reversed neuropathic pain (NP) symptoms, restored spatial memory integrity and decreased the expression of pTyr(1336)NR2B. Following postoperative administration of dextromethorphan, this study has demonstrated for the first time a concordance between behaviour, cognitive function and molecular events via pTyr(1336)NR2B for neuropathic pain alleviation. Confirmation of these findings in patients would constitute a major step forward in the treatment of neuropathic pain and in the improvement of cognitive function and quality of life.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Anti-NMDAR2B Antibody, phosphoTyr 1472, Chemicon®, from rabbit
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material