Merck
  • Development of a stability-indicating HPLC method of etifoxine with characterization of degradation products by LC-MS/TOF, 1H and 13C NMR.
Development of a stability-indicating HPLC method of etifoxine with characterization of degradation products by LC-MS/TOF, 1H and 13C NMR.
Journal of pharmaceutical and biomedical analysis (2014-08-15)
Nadia Djabrouhou, Moulay-Hassane Guermouche
ABSTRACT
This paper describes a new LC-MS/TOF method for the degradation products determination when Etifoxine (ETI) is submitted to different stress conditions. Chromatography is performed by using Kromasil C18 column (250mm×4.6mm, 5μm particle size). The selected mobile phase consists of formate buffer 0.02M, pH 3 and methanol (70/30, v/v). ETI is submitted to oxidative, acidic, basic, hydrolytic, thermal and UV light degradations. Detection is made at 254nm by photodiode array detector and mass spectrometry. A number of degradation products (DPs) called DPA, DPB, DPC and DPD are found depending on the stress; DPA with heat, DPA and DPB in acidic media or under UV-light; DPA, DPB and DPC under basic stress; DPA, DPB, DPC and DPD with oxidation. LC-MS/TOF is used to characterize the four DPs of ETI resulting from different stress conditions. (1)H and (13)C NMR are used to confirm the DP structures. The ETI fragmentation pathway is proposed. The method is validated with reference to International Conference on Harmonization guidelines and ETI are selectively determined in presence of its DPs, demonstrating its stability-indicating nature. Finally, for the validation step, specificity, linearity, accuracy and precision are determined for ETI and its DPs.
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%