Merck
  • Development of a biomimetic phospholipid vesicle-based permeation assay for the estimation of intestinal drug permeability.
Development of a biomimetic phospholipid vesicle-based permeation assay for the estimation of intestinal drug permeability.
Journal of pharmaceutical sciences (2014-03-26)
Elenaz Naderkhani, Johan Isaksson, Alexey Ryzhakov, Gøril Eide Flaten
ABSTRACT
Permeability is a crucial property of orally administered drugs. Therefore, in drug discovery, it is important to employ methods suitable for rapidly and reliably screening the permeability of large numbers of new drug candidates. The phospholipid vesicle-based permeation assay (PVPA), a model consisting of a tight layer of liposomes immobilized on a filter, offers potential advantages unmet by other methods and has been successfully used in permeability testing of novel active substances as well as formulations. In this study, the PVPA was developed into a more robust, biomimetic model by employing a lipid composition matching that of the intestinal permeation barrier and performing the experiments at the more biologically relevant pH 6.2. As expected, positively charged basic compounds demonstrated increased permeability through the negatively charged biomimetic barriers, and the degree of correct classification according to in vivo absorption was comparable between the original PVPA and the biomimetic PVPA. The biomimetic PVPA further proved to be tremendously more robust toward the presence of tensides compared with the original PVPA; this is a promising finding that renders the biomimetic PVPA an enhanced ability to estimate the permeability of poorly soluble compounds. Hence, the PVPA model developed in this study has evolved an important step forward.
Product Description
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Cimetidine
Supelco
Metronidazole, analytical standard
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Testosterone, European Pharmacopoeia (EP) Reference Standard
Cimetidine, European Pharmacopoeia (EP) Reference Standard
Timolol maleate, European Pharmacopoeia (EP) Reference Standard
Metronidazole, European Pharmacopoeia (EP) Reference Standard
USP
Cimetidine, United States Pharmacopeia (USP) Reference Standard
Supelco
Metronidazole, VETRANAL®, analytical standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen
Supelco
Diclofenac sodium salt, analytical standard
Diclofenac for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%