Merck
  • Description of the population structure and genetic diversity of tuberculosis in Estado de México, a low prevalence setting from Mexico.
Description of the population structure and genetic diversity of tuberculosis in Estado de México, a low prevalence setting from Mexico.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2014-09-27)
Roberto Zenteno-Cuevas, Fabiola Mendoza-Damián, Irving Cansino Muñoz, Leonor Enciso-Moreno, Lucia Monserrat Pérez-Navarro, Ma Dolores Ramírez-Hernández, Karen Vázquez-Medina, Lorena Widrobo-García, Michael Lauzardo, José Antonio Enciso-Moreno
ABSTRACT
In order to identify the genetic characteristics of the strains of mycobacteria circulating in the Estado de México, one of the states with the lowest prevalence of tuberculosis in Mexico, spoligotyping and 12-loci MIRU-VNTR typing were used to genotype tuberculosis clinical isolates. The average age of the 183 patients analyzed was 50 (± 17) years, drug resistance was noted in 57 (31%) and multidrug resistance in 22 (12%) individuals. The results from the isolates recovered showed that 80% were located in four major Euro-American lineages: Haarlem (17%), LAM (15%), T (20%) and X (29%). Other lineages found in lower proportions were: EAI, S, Beijing, West African, Turkey, Vole and Bovis. Eighteen isolates were orphans. Only 57 isolates were grouped in nine clusters and the SIT119 (X1) showed the highest number of members (23). The LAM lineage showed an increased risk for development of drug resistance (RR=4, IC: 95%: 1.05-14.2, p = 0.03). Despite the important prevalence of four major lineages found and the diversity of strains circulating in the population, we found the presence of one of the largest populations of isolates clustered to the X lineage in a setting from a Latin American country.
Product Description
Pyrazinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyrazinecarboxamide