Merck
Isolation and immortalization of MIP-GFP neurons from the hypothalamus.
Endocrinology (2014-03-13)
Zi Chen Wang, Michael B Wheeler, Denise D Belsham
ABSTRACT
The mouse insulin I promoter (MIP) construct was developed to eliminate the promoter activity detected with the rat insulin II promoter in specific hypothalamic neurons that may have unintended effects on glucose and energy homeostasis in transgenic models. Thus, the specificity of this novel construct must be validated prior to the widespread availability of derived Cre models. Although limited validation efforts have indicated a lack of MIP activity within neuronal tissue, the global immunohistochemical methodology used may not be specific enough to rule out the possibility of specific populations of neurons with MIP activity. To investigate possible MIP activity within the hypothalamus, primary hypothalamic isolates from MIP-green fluorescent protein reporter mice were analyzed after fluorescent-activated cell sorting. Primary hypothalamic neurons isolated from the MIP-green fluorescent protein mice were immortalized. Characterization detected the presence of hypothalamic neuropeptide Y (NPY) and agouti-related peptide, involved in the control of energy homeostasis, as well as confirmed insulin responsiveness in the cell lines. Moreover, because insulin was demonstrated to differentially regulate NPY expression within these MIP neurons, the promoter construct may be active in multiple hypothalamic NPY/agouti-related peptide subpopulations with unique physiological functions. MIP transgenic animals may therefore face similar limitations seen previously with rat insulin II promoter-based models.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, analytical standard
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%