Merck
  • A recombinant truncated surface immunogenic protein (tSip) plus adjuvant FIA confers active protection against Group B streptococcus infection in tilapia.
A recombinant truncated surface immunogenic protein (tSip) plus adjuvant FIA confers active protection against Group B streptococcus infection in tilapia.
Vaccine (2014-12-03)
Yang He, Kai-Yu Wang, Dan Xiao, De-Fang Chen, Lingyuan Huang, Tianqiang Liu, Jun Wang, Yi Geng, Er-Long Wang, Qian Yang
ABSTRACT
Tilapia is an important agricultural fish that has been plagued by Group B streptococcus (GBS) infections in recent years, some of them severe. It is well-known that surface immunogenicity protein (Sip) is an effective vaccine against GBS. Since Sip was not expressed in either E. coli BL21 or E. coli Rosetta, we removed the N-terminal signal peptide and LysM of the virus to produce purified truncated Sip (tSip(1)), which multiplied easily in an E. coli host. The antibody's ability to recognize and combine with GBS was determined by Western-blot and specific staining in vitro. The relative percentage of survival (RPS), antibody titers, bacterial recovery, and pathologic morphology were monitored in vivo to evaluate the immune effects. Freund's incomplete adjuvant (FIA) plus tSip and aluminum hydroxide gel (AH) plus tSip were also evaluated. It revealed that tSip mixed with FIA was an effective vaccine against GBS in tilapia, while AH is toxic to tilapia.
Product Description
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Supelco
Nitrilotriacetic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside