Merck
  • A novel quantitative body shape score for detecting association between obesity and hypertension in China.
A novel quantitative body shape score for detecting association between obesity and hypertension in China.
BMC public health (2015-01-18)
Shukang Wang, Yanxun Liu, Fangyu Li, Hongying Jia, Longjian Liu, Fuzhong Xue
ABSTRACT
Obesity is a major independent risk factor for chronic diseases such as hypertension and coronary diseases, it might not be only related to the amount of body fat but its distribution. The single body mass index (BMI), waist circumference (WC), waist to hip ratio (WHR) or waist to stature ratio (WSR) provides limited information on fat distribution, and the debate about which one is the best remained. On the other hand, the current classification of body shape is qualitative rather than quantitative, and only crudely measure fat distribution. Therefore, a synthetical index is highly desirable to quantify body shape. Based on the China Health and Nutrition Survey (CHNS) data, using Lohmäller PLSPM algorithm, six Partial Least Squares Path Models (PLSPMs) between the different obesity measurements and hypertension as well as two synthetical body shape scores (BSS1 by BMI/WC/Hip circumference, BSS2 by BMI/WC/WHR/WSR) were created. Simulation and real data analysis were conducted to assess their performance. Statistical simulation showed the proposed model was stable and powerful. Totally 15,172 (6,939 male and 8,233 female) participants aged from 18 to 87 years old were included. It indicated that age, height, weight, WC, WHR, WSR, SBP, DBP, the prevalence of hypertension and obesity were significantly sex-different. BMI, WC, WHR, WSR, Hip, BSS1 and BSS2 between hypertension and normotensive group are significantly different (p < 0.05). PLSPM method illustrated the biggest path coefficients (95% confidence interval, CI) were 0.220(0.196, 0.244) for male and 0.205(0.182, 0.228) for female in model of BSS1. The area under receiver-operating characteristic curve (AUC(95% CI)) of BSS1(0.839(0.831,0.847)) was significantly larger than that of BSS2(0.834(0.825,0.842)) as well as the four single indices for female, and similar trend can be found for male. BSS1 was an excellent measurement for quantifying body shape and detecting the association between body shape and hypertension.
Product Description
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dibutyl phthalate, 99%
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)