Merck
  • Development and validation of an UPLC-MS/MS method for the quantification of ethoxzolamide in blood, brain tissue, and bioequivalent buffers: applications to absorption, brain distribution, and pharmacokinetic studies.
Development and validation of an UPLC-MS/MS method for the quantification of ethoxzolamide in blood, brain tissue, and bioequivalent buffers: applications to absorption, brain distribution, and pharmacokinetic studies.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-24)
Song Gao, Jing Zhao, Taijun Yin, Yong Ma, Beibei Xu, Anthony N Moore, Pramod K Dash, Ming Hu
ABSTRACT
The purpose of this study is to develop and validate an UPLC-MS/MS method to quantify ethoxzolamide in plasma (EZ) and apply the method to absorption, brain distribution, as well as pharmacokinetic studies. A C₁₈ column was used with 0.1% of formic acid in acetonitrile and 0.1% of formic acid in water as the mobile phases to resolve EZ. The mass analysis was performed in a triple quadrupole mass spectrometer using multiple reaction monitoring (MRM) with positive scan mode. The results show that the linear range of EZ is 4.88-10,000.00 nM. The intra-day variance is less than 12.43% and the accuracy is between 88.88 and 108.00%. The inter-day variance is less than 12.87% and accuracy is between 89.27 and 115.89%. Protein precipitation was performed using methanol to extract EZ from plasma and brain tissues. Only 40 μL of plasma is needed for analysis due to the high sensitivity of this method, which could be completed in less than three minutes. This method was used to study the pharmacokinetics of EZ in SD rats, and the transport of EZ in Caco-2 and MDCK-MDR1 overexpressing cell culture models. Our data show that EZ is not a substrate for p-glycoprotein (P-gp) and its entry into the brain may not limited by the blood-brain barrier.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
(±)-Sulpiride
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sulpiride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard