Merck
  • Acute exercise ameliorates differences in insulin resistance between physically active and sedentary overweight adults.
Acute exercise ameliorates differences in insulin resistance between physically active and sedentary overweight adults.
Applied physiology, nutrition, and metabolism = Physiologie appliquee, nutrition et metabolisme (2014-04-30)
Rachael K Nelson, Jeffrey F Horowitz
ABSTRACT
Although regular exercise is associated with reduced cardiometabolic disease risk among overweight adults, it remains unclear whether much of the health benefits of exercise are derived from the most recent session(s) of exercise or if they are the result of adaptations stemming from weeks, months, or even years of training. The purpose of this study was to compare the effects of habitual and acute exercise on key markers of cardiometabolic disease risk in overweight adults. We compared insulin sensitivity index (ISI) using an oral glucose tolerance test, blood pressure (BP), blood lipids, and systemic inflammatory cytokines in 12 overweight to mildly obese adults (BMI: 27-34 kg/m(2)) who exercise regularly (EX; >2.5 h exercise per week) with a well-matched cohort of 12 nonexercisers (Non-EX). Baseline measurements in EX were performed exactly 3 days after exercise, whereas Non-EX remained sedentary. We repeated these measurements the day after a session of exercise in both groups. At baseline, ISI was significantly greater in EX versus Non-EX (3.1 ± 0.2 vs. 2.3 ± 0.2; p = 0.02), but BP, blood lipids, and plasma concentration of the systemic inflammatory cytokines we measured were not different between groups. Acute exercise increased ISI the next morning in Non-EX (2.3 ± 0.2 vs. 2.8 ± 0.3; p = 0.03) but not EX. As a result, ISI was similar between groups the morning after exercise. In summary, exercising regularly was accompanied by a persistent improvement in insulin sensitivity that lasted at least 3 days after exercise in overweight adults, but just one session of exercise increased insulin sensitivity among sedentary overweight adults to levels equivalent to the regular exercisers.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard