Merck
  • Quantification of intact human insulin-like growth factor-I in serum by nano-ultrahigh-performance liquid chromatography/tandem mass spectrometry.
Quantification of intact human insulin-like growth factor-I in serum by nano-ultrahigh-performance liquid chromatography/tandem mass spectrometry.
Rapid communications in mass spectrometry : RCM (2014-05-28)
Filipe Lopes, David A Cowan, Mario Thevis, Andreas Thomas, Mark C Parkin
ABSTRACT
Insulin-like growth factor-I is one of the biomarkers used to detect growth hormone administration prohibited in human sport. Current testing approaches for IGF-I rely on commercial immunoassays, which may change from time to time requiring complex revalidation. Mass spectrometry (MS)-based approaches often rely on enzymatically digesting the protein and measuring specific peptide concentrations. In order to reinforce the current available methodology for IGF-I testing, a reliable and equally sensitive MS method is required for the analysis of intact protein using small sample volumes (<25 μL). IGF-I was extracted from human serum samples by a simple protein precipitation procedure. Separation was achieved via nano-ultrahigh-performance liquid chromatography and MS analysis was conducted by nano-electrospray ionisation triple-quadrupole mass spectrometry in the selected reaction monitoring mode using a stable-isotope-labelled internal standard. A six-point calibration curve ranging from 50 to 1000 ng/mL of human IGF-I in rat serum was used to establish instrument response. The method provided a limit of quantification of 50 ng/mL, with intra- and inter-day precision ≤5% and intra- and inter-day accuracy ≥95%. A quantitative method was developed for the quantification of intact IGF-I in human serum samples. The data generated provided important information for the development of a new reference method for the growth hormone biomarker test and helped create a reliable system for monitoring peptide hormones in individual athletes, a possible extension to the athlete biological passport system. Nano-electrospray has here been shown to be sufficiently robust for routine use in an analytical laboratory, allowing for the analysis of minute sample volumes.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
IGF-II human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%