Merck
  • Neutralization of TNFSF10 ameliorates functional outcome in a murine model of Alzheimer's disease.
Neutralization of TNFSF10 ameliorates functional outcome in a murine model of Alzheimer's disease.
Brain : a journal of neurology (2014-12-05)
Giuseppina Cantarella, Giulia Di Benedetto, Daniela Puzzo, Lucia Privitera, Carla Loreto, Salvatore Saccone, Salvatore Giunta, Agostino Palmeri, Renato Bernardini
ABSTRACT
Alzheimer's disease is one of the most common causes of death worldwide, with poor treatment options. A tissue landmark of Alzheimer's disease is accumulation of the anomalous protein amyloid-β in specific brain areas. Whether inflammation is an effect of amyloid-β on the Alzheimer's disease brain, or rather it represents a cause for formation of amyloid plaques and intracellular tangles remains a subject of debate. TNFSF10, a proapoptotic cytokine of the TNF superfamily, is a mediator of amyloid-β neurotoxicity. Here, we demonstrate that blocking TNFSF10 by administration of a neutralizing monoclonal antibody could attenuate the amyloid-β-induced neurotoxicity in a triple transgenic mouse model of Alzheimer's disease (3xTg-AD). The effects of TNFSF10 neutralization on either cognitive parameters, as well as on the expression of TNFSF10, amyloid-β, inflammatory mediators and GFAP were studied in the hippocampus of 3xTg-AD mice. Treatment with the TNFSF10 neutralizing antibody resulted in dramatic improvement of cognitive parameters, as assessed by the Morris water maze test and the novel object recognition test. These results were correlated with decreased protein expression of TNFSF10, amyloid-β, inflammatory mediators and GFAP in the hippocampus. Finally, neutralization of TNFSF10 results in functional improvement and restrained immune/inflammatory response in the brain of 3xTg-AD mice in vivo. Thus, it is plausible to regard the TNFSF10 system as a potential target for efficacious treatment of amyloid-related disorders.
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized