Merck
  • Pharmacokinetics, bioavailability, and metabolism of Notoginsenoside Fc in rats by liquid chromatography/electrospray ionization tandem mass spectrometry.
Pharmacokinetics, bioavailability, and metabolism of Notoginsenoside Fc in rats by liquid chromatography/electrospray ionization tandem mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2015-03-17)
Chunyong He, Jia Li, Niusheng Xu, Rufeng Wang, Zeyun Li, Li Yang, Zhengtao Wang
ABSTRACT
Notoginsenoside Fc (NGFc) is a protopanaxadiol-type (PPD-type) saponin from Panax notoginseng, which has perfect anti-platelet aggregatory effect. However, its pharmacokinetics and metabolism in vivo remain unknown. In this study, a simple and sensitive liquid chromatography/electrospray ionization tandem mass spectrometry (LC-MS/MS) method was first developed for the determination of NGFc in rat plasma. After methanol-mediated protein precipitation, separation was achieved on a C18 column with MS detection operated in negative SRM mode at m/z 604.56→m/z 783.90 and m/z 799.93→m/z 637.64 for NGFc and IS, respectively. The assay was linear over the concentration range (r>0.995) with the LLOQ of 0.002μg/ml. The intra- and inter-day precisions (R.S.D.) were 2.45-12.36% and 3.67-14.22%, respectively; whereas accuracy ranged from (R.R.) 93.90% to 99.41%. The extraction recovery, stability, and matrix effect were within the acceptable limits. The validated LC-MS/MS method was successfully applied to the pre-clinical pharmacokinetic studies of NGFc in rat. After oral and intravenous administration, NGFc showed dose-independent pharmacokinetic behaviors with a t1/2 of >22h and its oral bioavailability was 0.10-0.14%. In addition, a total of 10 metabolites were detected and structurally characterized by UPLC-Q/TOF-MS technique, which suggested that deglycosylation was the major metabolic pathway for NGFc in rats.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ginsenoside Rg3, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Ginsenoside Rg3, analytical standard