Merck
  • Synergistic interaction of sumac and raspberry mixtures in their antioxidant capacities and selective cytotoxicity against cancerous cells.
Synergistic interaction of sumac and raspberry mixtures in their antioxidant capacities and selective cytotoxicity against cancerous cells.
Journal of medicinal food (2014-10-15)
Sunan Wang, Fan Zhu, Massimo F Marcone
ABSTRACT
Previous works on staghorn sumac (Rhus hirta) were mostly dedicated to its phytochemical profiles, antioxidant properties, and antidiabetic potentials. This study explored the potential of staghorn-sumac-derived functional ingredients for food and pharmacological applications. Sumac may have other biological functions, such as inhibitory effect on cancerous cells independent of its antioxidant properties. We characterized sumac and raspberry interactions, and their antioxidant capacities (ACs) and their inhibitory effect on both normal and cancerous cells. Mixing sumac and raspberry extracts yielded significantly higher ACs than the sum of sumac and raspberry as evaluated by three in vitro AC assays. However, the potential use of staghorn sumac as a natural source of dietary antioxidant supplement for oxidative-stress-related disorders might be challenged by its cytotoxicity in culturing normal cells. Remarkably, mixing sumac and raspberry showed maximal inhibition of the growth of both rat colon and human breast cancer cells with relatively low cytotoxicity toward normal rat colon and human breast epithelial cells, as compared with sumac or raspberry treatment alone. Sumac-derived products and their synergistic interactions with other food ingredients have great promise as functional food or nutraceutical products that would target cancer cells with minimal toxic effects to normal cells.
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Millipore
TDA Reagent, suitable for microbiology
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)