Merck
  • Housing temperature-induced stress drives therapeutic resistance in murine tumour models through β2-adrenergic receptor activation.
Housing temperature-induced stress drives therapeutic resistance in murine tumour models through β2-adrenergic receptor activation.
Nature communications (2015-03-11)
Jason W-L Eng, Chelsey B Reed, Kathleen M Kokolus, Rosemarie Pitoniak, Adam Utley, Mark J Bucsek, Wen Wee Ma, Elizabeth A Repasky, Bonnie L Hylander
ABSTRACT
Cancer research relies heavily on murine models for evaluating the anti-tumour efficacy of therapies. Here we show that the sensitivity of several pancreatic tumour models to cytotoxic therapies is significantly increased when mice are housed at a thermoneutral ambient temperature of 30 °C compared with the standard temperature of 22 °C. Further, we find that baseline levels of norepinephrine as well as the levels of several anti-apoptotic molecules are elevated in tumours from mice housed at 22 °C. The sensitivity of tumours to cytotoxic therapies is also enhanced by administering a β-adrenergic receptor antagonist to mice housed at 22 °C. These data demonstrate that standard housing causes a degree of cold stress sufficient to impact the signalling pathways related to tumour-cell survival and affect the outcome of pre-clinical experiments. Furthermore, these data highlight the significant role of host physiological factors in regulating the sensitivity of tumours to therapy.
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Corticosterone, VETRANAL®, analytical standard