Merck
  • The detrimental effects of acute hyperglycemia on myocardial glucose uptake.
The detrimental effects of acute hyperglycemia on myocardial glucose uptake.
Life sciences (2014-04-22)
Danzil Joseph, Charlene Kimar, Burger Symington, Robyn Milne, M Faadiel Essop
ABSTRACT
Although acute hyperglycemic (AHG) episodes are linked to lower glucose uptake, underlying mechanisms remain unclear. We hypothesized that AHG triggers reactive oxygen species (ROS) production and increases non-oxidative glucose pathway (NOGP) activation, i.e. stimulation of advanced glycation end products (AGE), polyol pathway (PP), hexosamine biosynthetic pathway (HBP), PKC; thereby decreasing cardiac glucose uptake. H9c2 cardiomyoblasts were exposed to 25 mM glucose for 24h vs. 5.5mM controls ± modulating agents during the last hour of glucose exposure: a) antioxidant #1 for mitochondrial ROS (250 μM 4-OHCA), b) antioxidant #2 for NADPH oxidase-generated ROS (100 μM DPI), c) NOGP inhibitors - 100 μM aminoguanidine (AGE), 5 μM chelerythrine (PKC); 40 μM DON (HBP); and 10 μM zopolrestat (PP). ROS levels (mitochondrial, intracellular) and glucose uptake were evaluated by flow cytometry. AHG elevated ROS, activated NOGPs and blunted glucose uptake. Transketolase activity (pentose phosphate pathway [PPP] marker) did not change. Respective 4-OHCA and DPI treatment blunted ROS production, diminished NOGP activation and normalized glucose uptake. NOGP inhibitory studies identified PKCβII as a key downstream player in lowering insulin-mediated glucose uptake. When we employed an agent (benfotiamine) known to shunt flux away from NOGPs (into PPP), it decreased ROS generation and NOGP activation, and restored glucose uptake under AHG conditions. This study demonstrates that AHG elicits maladaptive events that function in tandem to reduce glucose uptake, and that antioxidant treatment and/or attenuation of NOGP activation (PKC, polyol pathway) may limit the onset of insulin resistance.
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
S-Benzoylthiamine O-monophosphate
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Thiamine pyrophosphate, ≥95%
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Supelco
Thiamine pyrophosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology