Merck
  • Propionibacterium acnes in shoulder surgery: true infection, contamination, or commensal of the deep tissue?
Propionibacterium acnes in shoulder surgery: true infection, contamination, or commensal of the deep tissue?
Journal of shoulder and elbow surgery (2014-09-03)
Robert Hudek, Frank Sommer, Martina Kerwat, Ayman F Abdelkawi, Franziska Loos, Frank Gohlke
ABSTRACT
Propionibacterium acnes has been linked to chronic infections in shoulder surgery. Whether the bacterium is a contaminant or commensal of the deep tissue is unclear. We aimed to assess P. acnes in intraoperative samples of different tissue layers in patients undergoing first-time shoulder surgery. In 118 consecutive patients (mean age, 59.2 years; 75 men, 43 women), intraoperative samples were correlated to preoperative subacromial injection, the type of surgical approach, and gender. One skin, one superficial, one deep tissue, and one test sample were cultured for each patient. The cultures were positive for P. acnes in 36.4% (n = 43) of cases. Subacromial injection was not associated with bacterial growth rates (P = .88 for P. acnes; P = .20 for bacteria other than P. acnes; P = .85 for the anterolateral approach; P = .92 for the deltopectoral approach; P = .56 for men; P = .51 for women). Skin samples were positive for P. acnes in 8.5% (n = 10), superficial samples were positive in 7.6% (n = 9), deep samples were positive in 13.6% (n = 16), and both samples (superficial and deep) were positive in 15.3% (n = 18) of cases (P < .0001). P. acnes was detected in the anterolateral approach in 27.1% (n = 32) of cases and in the deltopectoral approach in 9.3% (n = 11) of cases (P = .01; relative risk, 1.93; 95% confidence interval, 1.08-3.43). Thirty-five of the P. acnes-positive patients were men (81.4%), and 8 patients were women (18.6%; P = .001; relative risk, 2.51; 95% confidence interval, 1.28-4.90). P. acnes was detected in more than one third of patients undergoing first-time shoulder surgery. Preoperative subacromial injection was not associated with bacterial growth. P. acnes was observed more frequently in the deep tissues than in the superficial tissues. The relative risk for obtaining a positive P. acnes culture was 2-fold greater for the anterolateral approach than for the deltopectoral approach, and the risk was 2.5-fold greater for men.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ampicillin
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard