Merck
  • Orostachys japonicus induces apoptosis and cell cycle arrest through the mitochondria-dependent apoptotic pathway in AGS human gastric cancer cells.
Orostachys japonicus induces apoptosis and cell cycle arrest through the mitochondria-dependent apoptotic pathway in AGS human gastric cancer cells.
International journal of oncology (2014-05-03)
Deok-Seon Ryu, Seon-Hee Kim, Ji-Hae Kwon, Dong-Seok Lee
ABSTRACT
We investigated the anticancer mechanisms of the ethylacetate (EtOAc) fraction from Orostachys japonicus in human gastric cancer (AGS) cells. Flow cytometric analysis revealed that the number of total apoptotic cells following treatment with the EtOAc fraction increased in a dose-dependent manner. In the cell cycle analyses, the EtOAc fraction increased the peak in the sub-G1, indicating apoptosis, and in the G₂/M phases in a dose-dependent manner. In the RT-PCR analysis, the expression of cyclin-dependent kinase 1 (CDK 1) and cyclin B1 decreased in a dose- and time-dependent manner. The results of western blotting revealed that the protein levels of p53, cytochrome c, and cleaved caspase-3, -8 and -9 proteins increased and those of B cell lymphoma-2 (bcl-2) and pro-caspase-3, -8 and -9 proteins decreased in a dose- and time-dependent manner, whereas the levels of bcl-2-associated x protein (bax) remained unchanged. Furthermore, the changes in the levels of pro-caspase-3, -8 and -9 and cleaved caspase-3, -8 and -9 were abolished by the pan-caspase inhibitor Z-VAD-FMK. In addition, phosphorylation of p38 and JNK increased in a time-dependent manner. These results, for the first time, provide an understanding of the potential anticancer activity of the O. japonicus, which functions through the induction of apoptosis and cell cycle arrest.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer