Merck
  • Improvement of transdermal delivery of sumatriptan succinate using a novel self-dissolving microneedle array fabricated from sodium hyaluronate in rats.
Improvement of transdermal delivery of sumatriptan succinate using a novel self-dissolving microneedle array fabricated from sodium hyaluronate in rats.
Biological & pharmaceutical bulletin (2015-03-12)
Dan Wu, Ying-shu Quan, Fumio Kamiyama, Kosuke Kusamori, Hidemasa Katsumi, Toshiyasu Sakane, Akira Yamamoto
ABSTRACT
The purpose of the present study was to develop an alternative transdermal formulation containing sumatriptan succinate (SS) for the treatment of migraine. Novel self-dissolving SS-loaded microneedle arrays (MNs) were fabricated from sodium hyaluronate and their efficacy for transdermal delivery of SS was characterized. The resulting MNs maintained their skin piercing abilities for at least 30 min after being placed at a high relative humidity of 75%. Rapid release of SS from the MNs was also observed in vitro. Optical coherence tomography images demonstrated that MNs were able to successfully pierce into rat skin without any bending or cracking, and needles were completely dissolved within 1 h. MNs significantly increased transepidermal water loss; however, skin barrier function gradually recovered to control levels within 24 h, in contrast to the skin damage observed after tape stripping treatment. These findings indicated that the micropores created by MNs quickly resealed, and that the skin damage was reversible. Furthermore, a dose-dependent plasma concentration of SS was obtained after transdermal delivery using SS-loaded MNs in rats. Absorption of SS delivered by MNs was similar to that observed after subcutaneous injection and was associated with high bioavailability (ca. 90%), which was much higher than that produced by oral administration. These findings suggested that application of SS-loaded MNs to the skin provided an effective alternative approach to enhance the transdermal delivery of SS without serious skin damage, and would be likely to improve patient compliance.
Product Description
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
Sigma-Aldrich
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
L-(+)-Tartaric acid, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
L-(+)-Tartaric acid, BioXtra
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Ammonium phosphate monobasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
L-(+)-Tartaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material