Merck
  • Assessment of comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry based methods for investigating 206 dioxin-like micropollutants in animal-derived food matrices.
Assessment of comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry based methods for investigating 206 dioxin-like micropollutants in animal-derived food matrices.
Journal of chromatography. A (2015-03-31)
Christelle Planche, Jérémy Ratel, Frédéric Mercier, Patrick Blinet, Laurent Debrauwer, Erwan Engel
ABSTRACT
This paper evaluates different multiresidue methods based on comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry (GC×GC-TOF/MS) to analyze dioxin-related micropollutants in complex food matrices. In a first step, the column sets Rtx-PCB/BPX-50 and Rtx-Dioxin2/BPX-50 were compared in terms of peak shape (width and symmetry) and resolution for the separation of polychlorinated biphenyls (PCBs) and polychlorinated dibenzo-p-dioxins/dibenzofurans (PCDD/Fs) in solvent. A satisfactory separation of 206 dioxin-related micropollutants including the 17 toxic PCDD/Fs was achieved in 75 min with the column set Rtx-Dioxin2/BPX-50. In a second time, the GC×GC-TOF/MS method was spread to the analysis of dioxin-related micropollutants in complex food matrices. An extraction procedure including accelerated solvent extraction (ASE), centrifugal evaporation and gel permeation chromatography (GPC) was optimized. Starting with meat as a model matrix, a micropollutant spiking method was then set up by comparing seven methods in terms of recoveries and reproducibility. The method combining immersion of the meat in a large volume of solvent containing micropollutants followed by homogenization by blender induced recoveries in the acceptable range of 70-130% and satisfactory standard deviations (≤10%) for most of the compounds studied. Limits of detection of the GC×GC-TOF/MS method ranged between 50 and 100 pg/g of spiked fresh meat for PCBs and between 65 and 227 pg/g for PCDD/Fs. Potential applications of this method are discussed.
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Toluene, analytical standard