Merck
  • Microwave-assisted synthesis of C-8 aryl and heteroaryl inosines and determination of their inhibitory activities against Plasmodium falciparum purine nucleoside phosphorylase.
Microwave-assisted synthesis of C-8 aryl and heteroaryl inosines and determination of their inhibitory activities against Plasmodium falciparum purine nucleoside phosphorylase.
European journal of medicinal chemistry (2014-06-16)
Alba Gigante, Eva-María Priego, Paula Sánchez-Carrasco, Luis Miguel Ruiz-Pérez, Johan Vande Voorde, María-José Camarasa, Jan Balzarini, Dolores González-Pacanowska, María-Jesús Pérez-Pérez
ABSTRACT
8-Arylinosines have been scarcely studied for therapeutic purposes, probably due to difficulties in their synthesis. The recently described direct arylation reaction at position 8 of purine nucleosides has been employed to synthesize a series of 8-aryl and 8-pyridylinosines. These compounds have been studied for hydrolytic stability and subjected to biological evaluation. Three compounds have shown a pronounced specific inhibition of Plasmodium falciparum-encoded purine nucleoside phosphorylase, an important target for antimalarial chemotherapy.
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard