Merck
The immune-related role of BRAF in melanoma.
Molecular oncology (2014-09-02)
Sara Tomei, Davide Bedognetti, Valeria De Giorgi, Michele Sommariva, Sara Civini, Jennifer Reinboth, Muna Al Hashmi, Maria Libera Ascierto, Qiuzhen Liu, Ben D Ayotte, Andrea Worschech, Lorenzo Uccellini, Paolo A Ascierto, David Stroncek, Giuseppe Palmieri, Lotfi Chouchane, Ena Wang, Francesco M Marincola
ABSTRACT
The existence of a dichotomy between immunologically active and quiescent tumor phenotypes has been recently recognized in several types of cancer. The activation of a Th1 type of immune signature has been shown to confer better prognosis and likelihood to respond to immunotherapy. However, whether such dichotomy depends on the genetic make-up of individual cancers is not known yet. BRAF and NRAS mutations are commonly acquired during melanoma progression. Here we explored the role of BRAF and NRAS mutations in influencing the immune phenotype based on a classification previously identified by our group. One-hundred-thirteen melanoma metastases underwent microarray analysis and BRAF and NRAS genotyping. Allele-specific PCR was also performed in order to exclude low-frequency mutations. Comparison between BRAF and NRAS mutant versus wild type samples identified mostly constituents or regulators of MAPK and related pathways. When testing gene lists discriminative of BRAF, NRAS and MAPK alterations, we found that 112 BRAF-specific transcripts were able to distinguish the two immune-related phenotypes already described in melanoma, with the poor phenotype associated mostly with BRAF mutation. Noteworthy, such association was stronger in samples displaying low BRAF mRNA expression. However, when testing NRAS mutations, we were not able to find the same association. This study suggests that BRAF mutation-related specific transcripts associate with a poor phenotype in melanoma and provide a nest for further investigation.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard