Merck
  • Preparation and application of sulfaguanidine-imprinted polymer on solid-phase extraction of pharmaceuticals from water.
Preparation and application of sulfaguanidine-imprinted polymer on solid-phase extraction of pharmaceuticals from water.
Talanta (2014-10-05)
Dragana Mutavdžić Pavlović, Korana Nikšić, Sara Livazović, Ivan Brnardić, Alojz Anžlovar
ABSTRACT
The molecularly imprinted polymers (MIPs) with sulfaguanidine as a template, methacrylic acid, 4-vinylpyridine, and 2-hydroxyethyl methacrylate as functional monomers, ethylene glycol dimethacrylate as a cross-linker and 2,2'-azobis-isobutyronitrile as an initiator have been prepared through the cross-link reaction of polymerization. Solid-phase extraction (SPE) procedure for the extraction of sulfaguanidine from water samples using the prepared MIPs and non-imprinted (NIPs) was evaluated. The best MIP in combination with commercial sorbents was applied for simultaneous extraction of eight pharmaceuticals. New SPE cartridges were prepared by combination of optimal produced MIP and Oasis HLB in 6 mL of polypropilene SPE reservoir. The developed method which includes new SPE cartridge (MIPMAA-Oasis HLB, 400mg/6 mL) and thin-layer chromatography was validated. The method provides a linear response over the concentration range of 0.5-150 μg/L, depending on the pharmaceutical with the correlation coefficients>0.9843 in all cases except for norfloxacin (0.9770) and penicillin G procaine (0.9801). Also, the method has revealed low limits of detection (0.25-20 μg/L), good precision (intra and inter-day), a relative standard deviation below 15% and recoveries above 95% for all eight pharmaceuticals. The developed method by using newly prepared SPE cartridge has been successfully applied to the analysis of production wastewater samples from pharmaceutical industry.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard