Merck
  • LC-MS/MS and GC-MS/MS measurement of plasma and urine di-paracetamol and 3-nitro-paracetamol: proof-of-concept studies on a novel human model of oxidative stress based on oral paracetamol administration.
LC-MS/MS and GC-MS/MS measurement of plasma and urine di-paracetamol and 3-nitro-paracetamol: proof-of-concept studies on a novel human model of oxidative stress based on oral paracetamol administration.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-04-29)
Arne Trettin, Darko Modun, Sanja Madunic, Jonatan Vukovic, Maja Radman, Sandor Batkai, Thomas Thum, Jens Jordan, Dimitrios Tsikas
ABSTRACT
Paracetamol (acetaminophen) is a widely used safe analgesic drug when administered at therapeutic doses. Given the chemical reactivity of its phenolic group towards electrophilic species, we assumed that detection of paracetamol metabolites distinctly different from its known phase I metabolite N-acetyl-p-benzoquinone imine (NAPQI) and the phase II glucuronic, sulfuric and mercapturic acids in biological samples upon oral administration of paracetamol (e.g., a 500-mg tablet) may represent a novel model of oxidative stress in humans. Such potential paracetamol metabolites are di-paracetamol and 3-nitro-paracetamol, in analogy to the well-investigated endogenous biomarkers di-tyrosine and 3-nitro-tyrosine. Di-paracetamol and 3-nitro-paracetamol are known to be formed both by enzymatic and non-enzymatic routes. In the present work we report on mouse and human pilot studies on the formation and appearance of di-paracetamol and 3-nitro-paracetamol in blood of mice intraperitoneally administered paracetamol, as well as in plasma and urine samples of healthy subjects who received a 500-mg paracetamol tablet or placebo. For the analysis of di-paracetamol and 3-nitro-paracetamol in plasma and urine samples, analytes were extracted by solvent extraction with ethyl acetate and subsequently analyzed by LC-MS/MS without and with derivatization with pentafluorobenzyl bromide. GC-MS/MS was used to detect 3-nitro-paracetamol and quantify paracetamol as pentafluorobenzyl derivatives. Our studies indicate that di-paracetamol and 3-nitro-paracetamol appear in plasma and urine when paracetamol is given orally to healthy humans at the therapeutic dosage of 5-7 mg/kg. The molar ratio of di-paracetamol to paracetamol in urine was determined to be 1:535 in the paracetamol group and 1:6844 in the placebo group; the molar ratio of 3-nitro-paracetamol to paracetamol in urine was determined to be 1:199 in the paracetamol group and 1:8657 in the placebo group. Our studies suggest that a fraction of circulating and excretory di-paracetamol and 3-nitro-paracetamol may be formed artefactually during sample workup including derivatization. Further studies based on the quantitative determination of di-paracetamol and 3-nitro-paracetamol in biological samples by LC-MS/MS and/or GC-MS/MS using stable-isotope labeled analogues as internal standards are warranted to test the utility of paracetamol as a probe of oxidative stress in animals and in humans in health and disease.
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzyl bromide, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard