Merck
  • A novel Amh-Treck transgenic mouse line allows toxin-dependent loss of supporting cells in gonads.
A novel Amh-Treck transgenic mouse line allows toxin-dependent loss of supporting cells in gonads.
Reproduction (Cambridge, England) (2014-09-13)
Mai Shinomura, Kasane Kishi, Ayako Tomita, Miyuri Kawasumi, Hiromi Kanezashi, Yoshiko Kuroda, Naoki Tsunekawa, Aisa Ozawa, Yoshimi Aiyama, Asuka Yoneda, Hitomi Suzuki, Michiko Saito, Jean-Yves Picard, Kenji Kohno, Masamichi Kurohmaru, Masami Kanai-Azuma, Yoshiakira Kanai
ABSTRACT
Cell ablation technology is useful for studying specific cell lineages in a developing organ in vivo. Herein, we established a novel anti-Müllerian hormone (AMH)-toxin receptor-mediated cell knockout (Treck) mouse line, in which the diphtheria toxin (DT) receptor was specifically activated in Sertoli and granulosa cells in postnatal testes and ovaries respectively. In the postnatal testes of Amh-Treck transgenic (Tg) male mice, DT injection induced a specific loss of the Sertoli cells in a dose-dependent manner, as well as the specific degeneration of granulosa cells in the primary and secondary follicles caused by DT injection in Tg females. In the testes with depletion of Sertoli cell, germ cells appeared to survive for only several days after DT treatment and rapidly underwent cell degeneration, which led to the accumulation of a large amount of cell debris within the seminiferous tubules by day 10 after DT treatment. Transplantation of exogenous healthy Sertoli cells following DT treatment rescued the germ cell loss in the transplantation sites of the seminiferous epithelia, leading to a partial recovery of the spermatogenesis. These results provide not only in vivo evidence of the crucial role of Sertoli cells in the maintenance of germ cells, but also show that the Amh-Treck Tg line is a useful in vivo model of the function of the supporting cell lineage in developing mammalian gonads.
Sigma-Aldrich
1-Decanethiol, 99%
Sigma-Aldrich
1-Decanethiol, 96%
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid